1-Phenyl-3-(trifluoromethyl)-2-pyrazolin-5-one 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Phenyl-3-(trifluoromethyl)-1H-pyrazol-5(4H)-one

B20478-14 B20478-03 B20478-06
CAAAAB20478-14EA 766.3 CAD
CAAAAB20478-14 CAAAB20478-03 CAAAB20478-06
1-Phenyl-3-(trifluoromethyl)-2-pyrazolin-5-one 98%
1-Phenyl-3-(trifluoromethyl)-2-pyrazolin-5-one

Purity: 98% CAS No.: 321-07-3 Molecular Formula: C10H7F3N2O Formula Weight: 228.18 Melting Point: 191-195° Beilstein Reference: 655085

Formula: C₁₀H₇F₃N₂O
MW: 228.17 g/mol
Melting Pt: 191…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00051655
CAS Number: 321-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR