4-Morpholinoaniline 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20292-06
B20292-14
CAAAAB20292-06EA
101.91
CAD
CAAAAB20292-06
CAAAAB20292-14
CAAAB20292-18
4-Morpholinoaniline 98+%
4-Morpholinoaniline
Purity: 98+%
CAS No.: 2524-67-6
Molecular Formula: C10H14N2O
Formula Weight: 178.24
Melting Point: 129-133°
Beilstein Reference: 150957
UN No.: 2811
MDL No.: MFCD00006169
Formula:
C₁₀H₁₄N₂O Melting Pt: 129…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006169 CAS Number: 2524-67-6 EINECS: 219-760-1 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...