Copper(II) trifluoromethanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Copper(II) trifluoromethanesulfonate, Copper(II) trifluoromethanesulphonate, Copper triflate

B20253-06 B20253-14 B20253-22
CAAAAB20253-06EA 141.25 CAD
CAAAAB20253-06 CAAAAB20253-14 CAAAAB20253-22
Copper(II) trifluoromethanesulfonate 98%
Copper (II) trifluoromethanesulphonate

Fieser: 7,76 8,126 10,110 14,102 19,112 20,122 21,149


Purity: 98% CAS No.: 34946-82-2 Molecular Formula: Cu(CF3SO3)2 Formula Weight: 361.68 Melting Point: dec. Beilstein Reference: 4028198 UN No.: 3261 MDL No.: MFCD00077492

Formula: C₂CuF₆O₆S₂
MW: 361.69 g/mol
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00077492
CAS Number: 34946-82-2
EINECS: 252-300-8
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR