trans-Aconitic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20087-14
B20087-22
CAAAAB20087-14EA
93.54
CAD
CAAAAB20087-14
CAAAAB20087-22
trans-Aconitic acid 98%
trans-Aconitic acid
Purity: 98%
CAS No.: 4023-65-8
Molecular Formula: C6H6O6
Formula Weight: 174.11
Odor: Odorless
Melting Point: ca 190° dec.
MDL No.: MFCD00002721
Formula:
C₆H₆O₆ Melting Pt: ca 190 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002721 CAS Number: 4023-65-8 EINECS: 223-688-6 Merck Index: 14,117 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...