N-Methylmorpholine-N-oxide 50% (w/w) in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Methylmorpholine N-Oxide, 4-Methylmorpholine 4-oxide
A19802-22
A19802-0B
A19802-30
CAAAAA19802-22EA
62.57
CAD
CAAAAA19802-22
CAAAA19802-0B
CAAAA19802-30
N-Methylmorpholine-N-oxide 50% (w/w) in aqueous solution
N-Methylmorpholine N-oxide
CAS No.: 7529-22-8
Molecular Formula: C5H11NO2
Formula Weight: 117.15
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 1.140
Refractive Index: 1.4220
Appearance: Colorless to yellow
Melting Point: -20°
Boiling Point: 118-119°
MDL No.: MFCD00005947
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol Boiling Pt: 118…119 °C Melting Pt: –20 °C Density: 1.140 Storage Temperature: Ambient |
MDL Number:
MFCD00005947 CAS Number: 7529-22-8 EINECS: 231-391-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...