(4-Fluorophenylthio)acetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-((4-Fluorophenyl)thio)acetic acid

A19658-03 A19658-06
CAAAA19658-03EA 74.1 CAD
CAAAA19658-03 CAAAA19658-06
(4-Fluorophenylthio)acetic acid 97%
(4-Fluorophenylthio)acetic acid

Purity: 97% CAS No.: 332-51-4 Molecular Formula: C8H7FO2S Formula Weight: 186.21 Melting Point: 76-79° MDL No.: MFCD00219825

Formula: C₈H₇FO₂S
MW: 186.21 g/mol
Melting Pt: 76…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00219825
CAS Number: 332-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR