2-Benzoxazolinone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19378-14
A19378-22
CAAAA19378-14EA
81.53
CAD
CAAAA19378-14
CAAAA19378-22
2-Benzoxazolinone 98%
2-Benzoxazolinone
Purity: 98%
CAS No.: 59-49-4
Molecular Formula: C7H5NO2
Formula Weight: 135.12
Flash Point: 160°(320°F)
Melting Point: 139-140°
Beilstein Reference: 119481
MDL No.: MFCD00005716
Formula:
C₇H₅NO₂ Melting Pt: 139…140 °C Flash Pt: 160 °C(320 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005716 CAS Number: 59-49-4 EINECS: 200-430-0 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...