Farnesol (mixture of isomers) 96%

Supplier: Thermo Scientific Chemicals

Synonyms: Farnesol, 3,7,11-Trimethyldodeca-2,6,10-trien-1-ol, TRANS,TRANS-3,7,11-TRIMETHYL-2,6,10-DODECATRIEN-1-OL, 2,6,10-Dodecatrien-1-ol, 3,7,11-trimethyl-, (2Z,6Z)-

A19316-06 A19316-14 A19316-22
CAAAAA19316-06EA 39.82 CAD
CAAAAA19316-06 CAAAA19316-14 CAAAA19316-22
Farnesol (mixture of isomers) 96%
Farnesol (mixture of isomers)

Applications: Induces apoptosis in cell cultures


Purity: 96% CAS No.: 4602-84-0 Molecular Formula: C15H26O Formula Weight: 222.37 Flash Point: 146°(295°F) Refractive Index: 1.4890 Odor: Pleasant Boiling Point: 159-160°/10mm Beilstein Reference: 1763926 Merck Reference: 14,3937 MDL No.: MFCD00002918

Formula: C₁₅H₂₆O
Boiling Pt: 159…160 °C/10 mm
Density: 0.887
Flash Pt: 146 °C(295 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002918
CAS Number: 4602-84-0
EINECS: 225-004-1
Merck Index: 14,3937

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR