Iminodibenzyl 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 10,11-Dihydro-5H-dibenzo[b,f]azepine

A19204-14 A19204-22
CAAAA19204-14EA 66.99 CAD
CAAAA19204-14 CAAAA19204-22
Iminodibenzyl 97%
Iminodibenzyl

Purity: 97% CAS No.: 494-19-9 Molecular Formula: C14H13N Formula Weight: 195.27 Odor: Odorless Melting Point: 105-108° Beilstein Reference: 152732 MDL No.: MFCD00005070

Formula: C₁₄H₁₃N
MW: 195.27 g/mol
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00005070
CAS Number: 494-19-9
EINECS: 207-787-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR