(R)-(-)-β-Aminophenethyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Danger

A19030-03 A19030-06 A19030-14
CAAAA19030-03EA 51.82 CAD
CAAAA19030-03 CAAAA19030-06 CAAAA19030-14
(R)-(-)-β-Aminophenethyl alcohol 98%
(R)-(-)-β-Aminophenethyl alcohol

Fieser: 15,256 17,279


Purity: 98% CAS No.: 56613-80-0 Molecular Formula: C8H11NO Formula Weight: 137.18 Melting Point: 76-78° Beilstein Reference: 2935848

Formula: C₈H₁₁NO
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00008062
CAS Number: 56613-80-0
EINECS: 260-287-5
UN: 3263
ADR: "8",III

Order Now

Specification Test Results

Optical rotation -30° (c=0.75 in 1N HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR