N,N-Dimethylglycine hydrochloride 99%

Supplier: Thermo Scientific Chemicals

A18892-06 A18892-14 A18892-22
CAAAA18892-06EA 57.99 CAD
CAAAA18892-06 CAAAA18892-14 CAAAA18892-22
N,N-Dimethylglycine hydrochloride 99%
N,N-Dimethylglycine hydrochloride

Purity: 99% CAS No.: 2491-06-7 Molecular Formula: (CH3)2NCH2CO2H.HCl Formula Weight: 139.58 Melting Point: 189-194° Beilstein Reference: 3910578 Merck Reference: 14,3244 MDL No.: MFCD00012610

Formula: C₄H₁₀ClNO₂
Melting Pt: 189…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00012610
CAS Number: 2491-06-7
EINECS: 219-648-2
Merck Index: 14,3244

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR