Acetylenedicarboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18736-06
A18736-14
A18736-22
CAAAA18736-06EA
54.35
CAD
CAAAA18736-06
CAAAA18736-14
CAAAA18736-22
Acetylenedicarboxylic acid 97%
Acetylenedicarboxylic acid
Fieser: 1,12
Purity: 97%
CAS No.: 142-45-0
Molecular Formula: HO2CC≡CCO2H
Formula Weight: 114.06
Storage Temperature: 2°C to 8°C
Flash Point: 131°(267°F)
Melting Point: ca 182° dec.
Beilstein Reference: 878357
UN No.: 2928
MDL No.: MFCD00004362
Formula:
C₄H₂O₄ Melting Pt: ca 182 °C (dec.) Flash Pt: 131 °C(267 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004362 CAS Number: 142-45-0 EINECS: 205-536-0 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...