1,3-Diphenylurea 98%

Supplier: Thermo Scientific Chemicals

A18720-14 A18720-22
CAAAA18720-14EA 53.72 CAD
CAAAA18720-14 CAAAA18720-22
1,3-Diphenylurea 98%
1,3-Diphenylurea

Purity: 98% CAS No.: 102-07-8 Molecular Formula: C6H5NHCONHC6H5 Formula Weight: 212.25 Melting Point: 240-244° Boiling Point: 260° dec. Merck Reference: 14,1785 MDL No.: MFCD00003017

Formula: C₁₃H₁₂N₂O
Boiling Pt: 260 °C (dec.)
Melting Pt: 240…244 °C
Density: 1.239
Storage Temperature: Ambient
MDL Number: MFCD00003017
CAS Number: 102-07-8
EINECS: 203-003-7
Merck Index: 14,1785

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR