Dipropyleneglycol (mixture of isomers) 99%
Supplier: Thermo Scientific Chemicals
A18716-0E
A18716-36
CAAAA18716-0EEA
180.12
CAD
CAAAA18716-0E
CAAAA18716-36
Dipropyleneglycol (mixture of isomers) 99%
Dipropyleneglycol (mixture of isomers)
Purity: 99%
CAS No.: 25265-71-8
Molecular Formula: HOC3H6OC3H6OH
Formula Weight: 134.18
Physical Form: Liquid
Density: 1.022
Refractive Index: 1.4410
Sensitivity: Hygroscopic
Appearance: Colorless
Odor: Odorless
Melting Point: -32°
Boiling Point: 231-235°
MDL No.: MFCD00051023
Formula:
C₆H₁₄O₃ Boiling Pt: 231…235 °C Melting Pt: –32 °C Density: 1.022 Storage Temperature: Ambient |
MDL Number:
MFCD00051023 CAS Number: 25265-71-8 EINECS: 246-770-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...