2-Chlorocyclohexanone 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with calcium carbonate/magnesium oxide
A18613-09
A18613-18
CAAAA18613-09EA
69.68
CAD
CAAAA18613-09
CAAAA18613-18
CAAAA18613-30
2-Chlorocyclohexanone 97% stabilized
2-Chlorocyclohexanone
Purity: 97%
CAS No.: 822-87-7
Molecular Formula: C6H9ClO
Formula Weight: 132.59
Storage Temperature: -30°C to -10°C
Flash Point: 82°(180°F)
Refractive Index: 1.4840
Melting Point: 22-24°
Boiling Point: 82-83°/10mm
Beilstein Reference: 774100
MDL No.: MFCD00001626
Formula:
C₆H₉ClO Boiling Pt: 82…83 °C/10 mm Melting Pt: 22…24 °C Density: 1.161 Flash Pt: 82 °C(180 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00001626 CAS Number: 822-87-7 EINECS: 212-505-5 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...