Copper(II) formate tetrahydrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18569-18
A18569-30
CAAAA18569-18EA
80.58
CAD
CAAAA18569-18
CAAAA18569-30
Copper(II) formate tetrahydrate 98%
Copper (II) formate tetrahydrate
Purity: 98%
CAS No.: 5893-61-8
Molecular Formula: (HCO2)2Cu.4H2O
Formula Weight: 225.65 (153.59anhy)
Merck Reference: 14,2639
UN No.: 3077
MDL No.: MFCD00167128
Formula:
C₂H₂CuO₄·4H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00167128 CAS Number: 5893-61-8 EINECS: 208-865-8 UN: 3077 ADR: "9",III Merck Index: 14,2639 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...