(±)-Methyl-2-chloropropionate 97%

Supplier: Thermo Scientific Chemicals
Warning

A18541-0B A18541-30
CAAAA18541-0BEA 413.96 CAD
CAAAA18541-0B CAAAA18541-30
(±)-Methyl-2-chloropropionate 97%
(±)-Methyl 2-chloropropionate

Purity: 97% CAS No.: 17639-93-9 Molecular Formula: C4H7ClO2 Formula Weight: 122.55 Flash Point: 38°(100°F) Refractive Index: 1.4170 Melting Point: -10° Boiling Point: 132-133° Beilstein Reference: 1421535 UN No.: 2933 MDL No.: MFCD00000863

Formula: C₄H₇ClO₂
Boiling Pt: 132…133 °C
Melting Pt: < –10 °C
Density: 1.140
Flash Pt: 38 °C(100 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000863
CAS Number: 17639-93-9
EINECS: 241-624-5
UN: 2933
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR