D-Panthenol 98+%
Supplier: Thermo Scientific Chemicals
A18499-22
A18499-36
CAAAA18499-22EA
87.85
CAD
CAAAA18499-22
CAAAA18499-36
D-Panthenol 98+%
D-Panthenol
Purity: 98+%
CAS No.: 81-13-0
Molecular Formula: C9H19NO4
Formula Weight: 205.26
Storage Temperature: 2°C to 8°C
Flash Point: 159°(318°F)
Refractive Index: 1.4990
Boiling Point: 118-120°/0.02m
Beilstein Reference: 1724947
Merck Reference: 14,2947
Optical Rotation: -30° (c=5 in water)
MDL No.: MFCD00065006
Formula:
C₉H₁₉NO₄ Boiling Pt: 118…120 °C/0.02 mm Density: 1.162 Flash Pt: 159 °C(318 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00065006 CAS Number: 81-13-0 EINECS: 201-327-3 Merck Index: 14,2947 |
Specification Test Results
Optical rotation | -30° (c=5 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...