Ammonium nickel (II) sulfate hexahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonium nickel sulphate hexahydrate, Nickel ammonium sulfate Ammonium disulfatonickelate, Ammonium nickel sulfate hexahydrate, Nickel ammonium sulfate hexahydrate, Nickel ammonium sulphate Ammonium disulphatonickelate, Nickel ammonium sulphate hexahydrate

A18441-22 A18441-36
CAAAAA18441-22EA 68.1 CAD
CAAAAA18441-22 CAAAAA18441-36
Ammonium nickel (II) sulfate hexahydrate 98%
Ammonium nickel (II) sulphate hexahydrate

Purity: 98% CAS No.: 7785-20-8 Molecular Formula: H8N2NiO8S2.6H2O Formula Weight: 394.99 (286.90anhy) Odor: Odorless Merck Reference: 14,6501 MDL No.: MFCD00150260

Formula: H₈N₂NiO₈S₂·6H₂O
MW: 394.99 g/mol
Density: 1.923
Storage Temperature: Ambient
MDL Number: MFCD00150260
CAS Number: 7785-20-8
EINECS: 239-793-5
UN: 3077
ADR: 9 III
Merck Index: 14,6501

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR