Galactitol 97%
Supplier: Thermo Scientific Chemicals
A18402-14
A18402-22
A18402-36
CAAAA18402-14EA
54.83
CAD
CAAAA18402-14
CAAAA18402-22
CAAAA18402-36
Galactitol 97%
Galactitol
Purity: 97%
CAS No.: 608-66-2
Molecular Formula: C6H14O6
Formula Weight: 182.17
Melting Point: 187-191°
Boiling Point: 275-280°/1mm
Merck Reference: 14,4332
MDL No.: MFCD00064288
Formula:
HOH₂C(CH(OH))₄CH₂OH Boiling Pt: 275…280 °C/1 mm Melting Pt: 187…191 °C Density: 1.47 Storage Temperature: Ambient |
MDL Number:
MFCD00064288 CAS Number: 608-66-2 EINECS: 210-165-2 Merck Index: 14,4332 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...