Succinyl chloride 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18389-18
A18389-30
CAAAA18389-18EA
90.38
CAD
CAAAA18389-18
CAAAA18389-30
Succinyl chloride 96%
Succinyl chloride
Purity: 96%
CAS No.: 543-20-4
Molecular Formula: ClC(O)CH2CH2C(O)Cl
Formula Weight: 154.98
Flash Point: 76°(168°F)
Refractive Index: 1.4710
Odor: Acrid
Melting Point: 16-17°
Boiling Point: 190°
Beilstein Reference: 507064
Merck Reference: 14,8873
UN No.: 3265
MDL No.: MFCD00000749
Formula:
C₄H₄Cl₂O₂ Boiling Pt: 190 °C Melting Pt: 16…17 °C Density: 1.407 Flash Pt: 76 °C(168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000749 CAS Number: 543-20-4 EINECS: 208-838-0 UN: 3265 ADR: "8",III Merck Index: 14,8873 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...