Nitrilotriacetic acid trisodium salt monohydrate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Nitrilotriacetic acid, trisodium salt, monohydrate

A18348-0E A18348-36
CAAAA18348-0EEA 319.16 CAD
CAAAA18348-0E CAAAA18348-36
Nitrilotriacetic acid trisodium salt monohydrate 98+%
Nitrilotriacetic acid trisodium salt monohydrate

Purity: 98+% CAS No.: 18662-53-8 Molecular Formula: N(CH2CO2Na)3.H2O Formula Weight: 275.12 (257.10anhy) Melting Point: 300° Beilstein Reference: 3774022 Merck Reference: 14,6579 MDL No.: MFCD00150789

Formula: C₆H₆NNa₃O₆·1H₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150789
CAS Number: 18662-53-8
EINECS: 205-355-7
Merck Index: 14,6579

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR