Rhodanine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Thioxo-4-thiazolidinone, 2-Thioxo-1,3-thiazolidin-4-one, 2-Thioxothiazolidin-4-one

A18293-14 A18293-22 A18293-36
CAAAA18293-14EA 78.53 CAD
CAAAA18293-14 CAAAA18293-22 CAAAA18293-36
Rhodanine 98+%
Rhodanine

Purity: 98+% CAS No.: 141-84-4 Molecular Formula: C3H3NOS2 Formula Weight: 133.19 Odor: Odorless Melting Point: 166-171° Beilstein Reference: 110701 Merck Reference: 14,8184 MDL No.: MFCD00005488

Formula: C₃H₃NOS₂
MW: 133.19 g/mol
Melting Pt: 166…171 °C
Density: 0.868
Storage Temperature: Ambient
MDL Number: MFCD00005488
CAS Number: 141-84-4
EINECS: 205-505-1
Merck Index: 14,8184

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR