Stearic acid hydrazide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Stearicacidhydrazide
A18291-09
A18291-18
A18291-30
CAAAA18291-09EA
58.93
CAD
CAAAA18291-09
CAAAA18291-18
CAAAA18291-30
Stearic acid hydrazide 97%
Stearic acid hydrazide
Purity: 97%
CAS No.: 4130-54-5
Molecular Formula: CH3(CH2)16CONHNH2
Formula Weight: 298.51
Melting Point: 114-116°
MDL No.: MFCD00014764
Formula:
C₁₈H₃₈N₂O MW: 298.51 g/mol Melting Pt: 114…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014764 CAS Number: 4130-54-5 EINECS: 223-946-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...