Perfluorodecahydronaphthalene (mixture of cis and trans isomers) 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Perfluoro(decahydronaphthalene), Perfluorodecalin, Perfluorodecahydronaphthalene

A18288-36 A18288-14 A18288-22
CAAAAA18288-36EA 1245.04 CAD
CAAAAA18288-36 CAAAA18288-14 CAAAA18288-22
Perfluorodecahydronaphthalene (mixture of cis and trans isomers) 95%
Perfluorodecahydronaphthalene (mixture of cis and trans isomers)

Purity: 95% CAS No.: 306-94-5 Molecular Formula: C10F18 Formula Weight: 462.10 Refractive Index: 1.3145 Melting Point: -10 to -7° Boiling Point: 141-142° Beilstein Reference: 2067113 Merck Reference: 14,4183 MDL No.: MFCD00010626

Formula: C₁₀F₁₈
MW: 462.08 g/mol
Boiling Pt: 141…142 °C
Melting Pt: –10 to –7 °C
Density: 1.940
Storage Temperature: Ambient
MDL Number: MFCD00010626
CAS Number: 306-94-5
EINECS: 206-192-4
UN: 1993
ADR: "3",III
Merck Index: 14,4183

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR