1-Naphthyl phosphate monosodium salt monohydrate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Naphthylphosphoric Acid Monosodium Salt Monohydrate, Monosodium 1-Naphthyl Phosphate Monohydrate, Sodium 1-Naphthyl Hydrogen Phosphate Monohydrate, Phosphoric Acid 1-Naphthyl Ester Monosodium Salt Monohydrate
A18254-22
A18254-06
A18254-14
CAAAAA18254-22EA
1594.22
CAD
CAAAAA18254-22
CAAAA18254-06
CAAAA18254-14
1-Naphthyl phosphate monosodium salt monohydrate 98+%
1-Naphthyl phosphate monosodium salt monohydrate
Purity: 98+%
CAS No.: 81012-89-7
Molecular Formula: C10H8NaO4P.H2O
Formula Weight: 264.16 (246.14anhy)
Storage Temperature: 2°C to 8°C
Melting Point: 186-191°
MDL No.: MFCD00150615
Formula:
C₁₀H₈NaO₄P·1H₂O MW: 264.15 g/mol Melting Pt: 186…191 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00150615 CAS Number: 81012-89-7 EINECS: 220-171-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...