(S)-(-)-4-Benzyl-2-oxazolidinone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18236-06
A18236-14
CAAAAA18236-03EA
97.33
CAD
CAAAAA18236-03
CAAAAA18236-06
CAAAAA18236-14
(S)-(-)-4-Benzyl-2-oxazolidinone 99%
(S)-(-)-4-Benzyl-2-oxazolidinone
Fieser: 17,247
Purity: 99%
CAS No.: 90719-32-7
Molecular Formula: C10H11NO2
Formula Weight: 177.20
Melting Point: 87-89°
Beilstein Reference: 3649667
Formula:
C₁₀H₁₁NO₂ Melting Pt: 87…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064496 CAS Number: 90719-32-7 |
Specification Test Results
Optical rotation | -64° (c=1 in chloroform) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...