DL-Ornithine hydrochloride 99%
Supplier: Thermo Scientific Chemicals
A18173-18
A18173-06
A18173-14
CAAAAA18173-18EA
232.26
CAD
CAAAAA18173-18
CAAAA18173-06
CAAAA18173-14
DL-Ornithine hydrochloride 99%
DL-Ornithine hydrochloride
Purity: 99%
CAS No.: 1069-31-4
Molecular Formula: C5H12N2O2.HCl
Formula Weight: 168.62
Melting Point: ca 225° dec.
Beilstein Reference: 4153338
Formula:
C₅H₁₂N₂O₂·HCl Melting Pt: ∼ 225 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00065398 CAS Number: 1069-31-4 EINECS: 213-956-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...