Cacodylic acid sodium salt trihydrate ≥98%, ee 98%

Supplier: Thermo Scientific Chemicals
Danger

A18139-14 A18139-22 A18139-36
CAAAA18139-14EA 140.94 CAD
CAAAA18139-14 CAAAA18139-22 CAAAA18139-36
Cacodylic acid sodium salt trihydrate ≥98%, ee 98%
Cacodylic acid sodium salt trihydrate

An integral component of terminal deoxynucleotidyl transferase buffer; also useful in electron microscopy as a bactericide buffer


Purity: 98+% CAS No.: 6131-99-3 Molecular Formula: C2H6AsNaO2.3H2O Formula Weight: 214.03 (159.98anhy) Beilstein Reference: 3702348 Merck Reference: 14,8595 MDL No.: MFCD00149079


An integral component of terminal deoxynucleotidyl transferase buffer; also useful in electron microscopy as a bactericide buffer

Formula: (CH₃)₂As(O)ONa·3H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149079
CAS Number: 6131-99-3
EINECS: 204-708-2
UN: 1688
ADR: 6.1,II
Merck Index: 14,8595

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR