DL-Menthol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18098-0B
A18098-18
A18098-22
A18098-30
CAAAAA18098-0BEA
388.68
CAD
CAAAAA18098-0B
CAAAAA18098-18
CAAAAA18098-22
CAAAAA18098-30
DL-Menthol 98+%
DL-Menthol
Has local anesthetic and counter-irritant effects; a weak kappa-opioid receptor agonist
Purity: 98+%
CAS No.: 89-78-1
Molecular Formula: C10H20O
Formula Weight: 156.27
Flash Point: 93°(199°F)
Refractive Index: 1.4615
Odor: Sweet
Melting Point: 33-36°
Boiling Point: 212-216°
Beilstein Reference: 3194263
Merck Reference: 14,5837
MDL No.: MFCD00001484
Formula:
C₁₀H₂₀O Boiling Pt: 212…216 °C Melting Pt: 33…36 °C Density: 0.890 Flash Pt: 93 °C(199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001484 CAS Number: 89-78-1 EINECS: 201-939-0 Merck Index: 14,5837 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...