2-Hydroxypropyl methacrylate (mixture of isomers) 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) ∼0.02%
A17951-0J
A17951-AK
A17951-AU
CAAAAA17951-0JEA
786.84
CAD
CAAAAA17951-0J
CAAAAA17951-AK
CAAAAA17951-AU
2-Hydroxypropyl methacrylate (mixture of isomers) 98% stabilized
2-Hydroxypropyl methacrylate (mixture of isomers, methacrylic acid, monoester with propane-1,2-diol)
Purity: 98%
CAS No.: 27813-02-1
Molecular Formula: CH2=C(CH3)CO2CH2CH(OH)CH3
Formula Weight: 144.17
Storage Temperature: 2°C to 8°C
Flash Point: 97°(207°F)
Refractive Index: 1.4480
Melting Point: 70°/1mm
Boiling Point: 205-209°
MDL No.: MFCD00004536
Formula:
C₇H₁₂O₃ Boiling Pt: 205…209 °C Melting Pt: 70 °C/1mm Density: 1.029 Flash Pt: 97 °C(207 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004536 CAS Number: 27813-02-1 EINECS: 248-666-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...