Diphenyliodonium hexafluorophosphate 98%

Supplier: Thermo Scientific Chemicals
Danger

A17934-09 A17934-18 A17934-30
CAAAAA17934-09EA 186.44 CAD
CAAAAA17934-09 CAAAAA17934-18 CAAAAA17934-30
Diphenyliodonium hexafluorophosphate 98%
Diphenyliodonium hexafluorophosphate

Purity: 98% CAS No.: 58109-40-3 Molecular Formula: C12H10F6IP Formula Weight: 426.08 Melting Point: 139-144° UN No.: 2811 MDL No.: MFCD00061398

Formula: C₁₂H₁₀F₆IP
MW: 426.08 g/mol
Melting Pt: 139…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00061398
CAS Number: 58109-40-3
EINECS: 261-134-5
UN: 1759
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR