Tetraethyl orthocarbonate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17835-22
A17835-14
CAAAAA17835-22EA
820.02
CAD
CAAAAA17835-22
CAAAA17835-14
Tetraethyl orthocarbonate 98+%
Tetraethyl orthocarbonate
Fieser: 1,1138
Purity: 98+%
CAS No.: 78-09-1
Molecular Formula: C9H20O4
Formula Weight: 192.26
Storage Temperature: 2°C to 8°C
Flash Point: 52°(125°F)
Refractive Index: 1.3930
Boiling Point: 158-161°
Beilstein Reference: 1747401
UN No.: 3272
MDL No.: MFCD00009221
Formula:
C₉H₂₀O₄ Boiling Pt: 158…161 °C Density: 0.917 Flash Pt: 52 °C (125 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009221 CAS Number: 78-09-1 EINECS: 201-082-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...