3-(Trimethoxysilyl)propyl methacrylate 97%
Supplier: Thermo Scientific Chemicals
A17714-22
A17714-36
CAAAAA17714-22EA
71.42
CAD
CAAAAA17714-22
CAAAAA17714-36
3-(Trimethoxysilyl)propyl methacrylate 97%
3-(Trimethoxysilyl)propyl methacrylate
Purity: 97%
CAS No.: 2530-85-0
Molecular Formula: C10H20O5Si
Formula Weight: 248.35
Flash Point: 92°(197°F)
Refractive Index: 1.431
Boiling Point: 190°
Beilstein Reference: 1952435
MDL No.: MFCD00008593
Formula:
C₁₀H₂₀O₅Si Boiling Pt: 190 °C Density: 1.045 Flash Pt: 92 °C(197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008593 CAS Number: 2530-85-0 EINECS: 219-785-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...