2-Imidazolidinone hemihydrate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Imidazolidone hemihydrate
A17666-0B
A17666-30
CAAAA17666-0BEA
150.1
CAD
CAAAA17666-0B
CAAAA17666-30
2-Imidazolidinone hemihydrate 98+%
2-Imidazolidinone hemihydrate
Purity: 98+%
CAS No.: 121325-67-5
Molecular Formula: C3H6N2O.0.5H2O
Formula Weight: 95.10 (86.09anhy)
Melting Point: 55°
MDL No.: MFCD02662355
Formula:
C₃H₆N₂O·0.5H₂O MW: 95.10 (86.09 anhy) Melting Pt: 55 °C Storage Temperature: Ambient |
MDL Number:
MFCD02662355 CAS Number: 121325-67-5 EINECS: 204-436-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...