Menthone (mixture of isomers) 98%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Menthone, 2-Isopropyl-5-Methylcyclohexanone
A17665-0B
A17665-18
A17665-30
CAAAA17665-0BEA
657.28
CAD
CAAAA17665-0B
CAAAA17665-18
CAAAA17665-30
Menthone (mixture of isomers) 98%
Menthone (mixture of isomers)
Purity: 98%
CAS No.: 10458-14-7
Molecular Formula: C10H18O
Formula Weight: 154.25
Flash Point: 72°(162°F)
Boiling Point: ca 207°
MDL No.: MFCD00062998
Formula:
C₁₀H₁₈O Boiling Pt: ca 207 °C Density: 0.894 Flash Pt: 72 °C(162 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00062998 CAS Number: 10458-14-7 EINECS: 233-944-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...