3,6-Dithia-1,8-octanediol 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,2-(Ethane-1,2-diylbis(sulfanediyl))diethanol, 1,8-Dihydroxy-3,6-Dithiaoctane, 2,2-(Ethane-1,2-diylbis(sulphanediyl))diethanol

A17654-09 A17654-18 A17654-30
CAAAA17654-09EA 85.16 CAD
CAAAA17654-09 CAAAA17654-18 CAAAA17654-30
3,6-Dithia-1,8-octanediol 97%
3,6-Dithia-1,8-octanediol

Purity: 97% CAS No.: 5244-34-8 Molecular Formula: HOCH2CH2SCH2CH2SCH2CH2OH Formula Weight: 182.31 Melting Point: 63-67° Boiling Point: 170°/0.8mm MDL No.: MFCD00002911

Formula: C₆H₁₄O₂S₂
MW: 182.31 g/mol
Boiling Pt: 170 °C/0.8 mm
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00002911
CAS Number: 5244-34-8
EINECS: 226-045-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR