L(-)-Carnitine ≥99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate

A17618-09 A17618-18
CAAAA17618-09EA 150.73 CAD
CAAAA17618-09 CAAAA17618-18 CAAAA17618-30
L(-)-Carnitine ≥99%
L(-)-Carnitine

Purity: 99+% CAS No.: 541-15-1 Molecular Formula: C7H15NO3 Formula Weight: 161.20 Melting Point: ca 190° dec. Beilstein Reference: 4292315 Merck Reference: 14,1849 MDL No.: MFCD00038747

Formula: C₇H₁₅NO₃
MW: 161.20 g/mol
Melting Pt: ∼ 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00038747
CAS Number: 541-15-1
EINECS: 208-768-0
Merck Index: 14,1849

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR