Magnesium trifluoromethanesulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A17591-06
A17591-14
A17591-22
CAAAA17591-06EA
52.46
CAD
CAAAA17591-06
CAAAA17591-14
CAAAA17591-22
Magnesium trifluoromethanesulfonate 98%
Magnesium trifluoromethanesulphonate
Purity: 98%
CAS No.: 60871-83-2
Molecular Formula: C2F6MgO6S2
Formula Weight: 322.43
Melting Point:
Formula:
C₂F₆MgO₆S₂ Melting Pt: >310 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042601 CAS Number: 60871-83-2 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...