Creatine (N-amidinosarcosine), anhydrous 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(1-Methylguanidino)acetic acid, N-Amidinosarcosine, Creatine

A17477-14 A17477-22
CAAAA17477-14EA 71.8 CAD
CAAAA17477-14 CAAAA17477-22
Creatine (N-amidinosarcosine), anhydrous 98%
Creatine (N-Amidinosarcosine)

Purity: 98% CAS No.: 57-00-1 Molecular Formula: C4H9N3O2 Formula Weight: 131.13 Physical Form: Powder Density: 1.33 Appearance: White Melting Point: ca 290° dec. Beilstein Reference: 907175 Merck Reference: 14,2568 MDL No.: MFCD00004282

Formula: C₄H₉N₃O₂
MW: 131.13 g/mol
Melting Pt: ∼ 290 °C (dec.)
Density: 1.33
Storage Temperature: Ambient
MDL Number: MFCD00004282
CAS Number: 57-00-1
EINECS: 200-306-6
Merck Index: 14,2568

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR