5-(4-(Dimethylamino)benzylidene)rhodanine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Dimethylaminobenzalrhodanine, 5-(4-Dimethylaminobenzylidene)-2-thioxo-4-thiazolidinone, p-Dimethylaminobenzalrhodanine, 5-(4-(Dimethylamino)benzylidene)-2-mercaptothiazol-4(5H)-one, p-(Dimethylamino)benzal-5-rhodanine, (5E)-5-[4-(Dimethylamino)benzylidene]-2-mercapto-1,3-thiazol-4(5H)-one

A17152-06 A17152-14
CAAAAA17152-06EA 81.84 CAD
CAAAAA17152-06 CAAAAA17152-14
5-(4-(Dimethylamino)benzylidene)rhodanine 98%
5-(4-(Dimethylamino)benzylidene)rhodanine

Purity: 98% CAS No.: 536-17-4 Molecular Formula: C12H12N2OS2 Formula Weight: 264.37 Odor: Weak Melting Point: 275-280° Beilstein Reference: 189065 Merck Reference: 14,3231 MDL No.: MFCD00064857

Formula: C₁₂H₁₂N₂OS₂
MW: 264.37 g/mol
Melting Pt: 275…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00064857
CAS Number: 536-17-4
EINECS: 208-625-2
Merck Index: 14,3231

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR