4-Quinazolone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxyquinazoline, Quinazolin-4(3H)-one

A17129-09 A17129-18
CAAAA17129-09EA 67.78 CAD
CAAAA17129-09 CAAAA17129-18
4-Quinazolone 98%
4-Quinazolone

Purity: 98% CAS No.: 491-36-1 Molecular Formula: C8H6N2O Formula Weight: 146.15 Odor: Odorless Melting Point: 216-219° Beilstein Reference: 118473 MDL No.: MFCD00511302

Formula: C₈H₆N₂O
MW: 146.15 g/mol
Melting Pt: 216…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00511302
CAS Number: 491-36-1
EINECS: 207-735-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR