4-Methoxy-3-nitrobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A17081-06
A17081-14
CAAAA17081-03EA
57.69
CAD
CAAAA17081-03
CAAAA17081-06
CAAAA17081-14
4-Methoxy-3-nitrobenzaldehyde 98%
4-Methoxy-3-nitrobenzaldehyde
Purity: 98%
CAS No.: 31680-08-7
Molecular Formula: C8H7NO4
Formula Weight: 181.15
Melting Point: 97-100°
MDL No.: MFCD00126498
Formula:
C₈H₇NO₄ Melting Pt: 97…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00126498 CAS Number: 31680-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...