Tetraethylene glycol monomethyl ether 98%

Supplier: Thermo Scientific Chemicals

Synonyms: mPEG4-OH, 2,5,8,11-Tetraoxatridecan-13-ol

A17021-06 A17021-14
CAAAA17021-06EA 114.87 CAD
CAAAA17021-06 CAAAA17021-14
Tetraethylene glycol monomethyl ether 98%
Tetraethylene glycol monomethyl ether

Purity: 98% CAS No.: 23783-42-8 Molecular Formula: H(OCH2CH2)4OCH3 Formula Weight: 208.25 Refractive Index: 1.4450 Boiling Point: 158-160°/5mm MDL No.: MFCD00041756

Formula: C₉H₂₀O₅
MW: 208.25 g/mol
Boiling Pt: 158…160 °C (5 mmHg)
Density: 1.07
Storage Temperature: Ambient
MDL Number: MFCD00041756
CAS Number: 23783-42-8
EINECS: 245-883-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR