Ammonium bismuth citrate Bi 48-52%, (max. 2% H₂O)

Supplier: Thermo Scientific Chemicals
Warning

A17003-0B A17003-18 A17003-30
CAAAA17003-0BEA 1044.38 CAD
CAAAA17003-0B CAAAA17003-18 CAAAA17003-30
Ammonium bismuth citrate Bi 48-52%, (max. 2% H₂O)
Ammonium bismuth citrate

Bi 50-52%


CAS No.: 25530-63-6 Molecular Formula: C6H10BiNO8 Formula Weight: 433.12 MDL No.: MFCD00036420

Formula: C₆H₁₁BiNO₇
Storage Temperature: Ambient
MDL Number: MFCD00036420
CAS Number: 25530-63-6
EINECS: 247-078-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR