N-Methylpropylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16903-03
A16903-06
A16903-14
CAAAAA16903-03EA
53.4
CAD
CAAAAA16903-03
CAAAAA16903-06
CAAAAA16903-14
N-Methylpropylamine 97%
N-Methylpropylamine
Purity: 97%
CAS No.: 627-35-0
Molecular Formula: CH3NHCH2CH2CH3
Formula Weight: 73.14
Flash Point: -1.1°(30°F)
Odor: Amine-like
Boiling Point: 62-64°
UN No.: 2733
MDL No.: MFCD00009361
Formula:
C₄H₁₁N Boiling Pt: 62…64 °C Density: 0.713 Flash Pt: –1.1 °C (30 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009361 CAS Number: 627-35-0 UN: 2733 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...