N,N'-Dimethyl-1,3-propanediamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N1,N3-Dimethylpropane-1,3-diamine, N,N′-Dimethyl-1,3-propanediamine

A16901-06 A16901-14
CAAAA16901-06EA 125.61 CAD
CAAAA16901-06 CAAAA16901-14
N,N'-Dimethyl-1,3-propanediamine 97%
N,N'-Dimethyl-1,3-propanediamine

Purity: 97% CAS No.: 111-33-1 Molecular Formula: CH3NH(CH2)3NHCH3 Formula Weight: 102.18 Flash Point: 32°(90°F) Odor: Amine-like Boiling Point: 143-146° UN No.: 2733 MDL No.: MFCD00008292

Formula: C₅H₁₄N₂
MW: 102.18 g/mol
Boiling Pt: 143…146 °C
Density: 0.817
Flash Pt: 32 °C (90 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008292
CAS Number: 111-33-1
EINECS: 203-859-1
UN: 2733
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR