3-Methylcrotonic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

A16865-14 A16865-22 A16865-36
CAAAAA16865-14EA 49.45 CAD
CAAAAA16865-14 CAAAA16865-22 CAAAA16865-36
3-Methylcrotonic acid 98%
3-Methylcrotonic acid

Purity: 98% CAS No.: 541-47-9 Molecular Formula: C5H8O2 Formula Weight: 100.12 Flash Point: 93°(199°F) Melting Point: 66-68° Boiling Point: 198-200° Beilstein Reference: 1720305 UN No.: 3261 MDL No.: MFCD00004366

Formula: C₅H₈O₂
Boiling Pt: 198…200 °C
Melting Pt: 66…68 °C
Density: 0.969
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004366
CAS Number: 541-47-9
EINECS: 208-782-7
UN: 3261
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR