2,2':5',2''-Terthiophene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16748-06
A16748-MD
A16748-03
CAAAAA16748-06EA
1036.48
CAD
CAAAAA16748-06
CAAAAA16748-MD
CAAAA16748-03
2,2':5',2''-Terthiophene 99%
2,2':5',2''-Terthiophene
Purity: 99%
CAS No.: 1081-34-1
Molecular Formula: C12H8S3
Formula Weight: 248.39
Melting Point: 89-94°
Boiling Point: 160°/0.1mm
Merck Reference: 14,9174
MDL No.: MFCD00012167
Formula:
C₁₂H₈S₃ Boiling Pt: 160 °C/0.1 mm Melting Pt: 89…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012167 CAS Number: 1081-34-1 Merck Index: 14,9174 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...