Diphenylboric acid-2-aminoethyl ester (2-APB) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A16606-06
A16606-14
CAAAA16606-06EA
123.24
CAD
CAAAA16606-06
CAAAA16606-14
Diphenylboric acid-2-aminoethyl ester (2-APB) 98%
Diphenylboric acid 2-aminoethyl ester (2-APB)
Purity: 98%
CAS No.: 524-95-8
Molecular Formula: C14H16BNO
Formula Weight: 225.10
Melting Point: 187-190°
Beilstein Reference: 2942273
Formula:
C₁₄H₁₆BNO Melting Pt: 187…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014823 CAS Number: 524-95-8 EINECS: 208-366-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...